Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113235.1 | NP_001106706.1 | 17 - 18 | 2028 | 140 | |
XM_023653807.1 | XP_023509575.1 | 2060 | 140 | |||
XM_023653808.1 | XP_023509576.1 | 1884 | 140 | |||
XM_023653810.1 | XP_023509578.1 | 1892 | 140 | |||
XM_023653811.1 | XP_023509579.1 | 2042 | 140 | |||
XM_023653812.1 | XP_023509580.1 | 2042 | 140 | |||
XM_023653813.1 | XP_023509581.1 | 2009 | 140 | |||
XM_023653814.1 | XP_023509582.1 | 2009 | 140 | |||
XM_023653815.1 | XP_023509583.1 | 1973 | 140 | |||
XM_023653816.1 | XP_023509584.1 | 1973 | 140 | |||
XM_023653817.1 | XP_023509585.1 | 1955 | 140 | |||
XM_023653818.1 | XP_023509586.1 | 1955 | 140 | |||
XM_023653819.1 | XP_023509587.1 | 1922 | 140 | |||
XM_023653821.1 | XP_023509589.1 | 1922 | 140 | |||
XM_023653822.1 | XP_023509590.1 | 1581 | 140 | |||
XM_023653823.1 | XP_023509591.1 | 1640 | 140 | |||
XM_023653824.1 | XP_023509592.1 | 2060 | 140 | |||
GenBank mRNA | AB371895.1 | - | 17 - 18 | 2028 | 140 | |
AB371896.1 | - | 16 - 17 | 1941 | 140 | ||
AB371897.1 | - | 16 - 17 | 1923 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |