Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113235.1 | NP_001106706.1 | 4 - 5 | 658 | 123 | |
XM_023653807.1 | XP_023509575.1 | 690 | 123 | |||
XM_023653808.1 | XP_023509576.1 | 514 | 123 | |||
XM_023653810.1 | XP_023509578.1 | 522 | 123 | |||
XM_023653811.1 | XP_023509579.1 | 690 | 123 | |||
XM_023653812.1 | XP_023509580.1 | 690 | 123 | |||
XM_023653813.1 | XP_023509581.1 | 690 | 123 | |||
XM_023653814.1 | XP_023509582.1 | 690 | 123 | |||
XM_023653815.1 | XP_023509583.1 | 690 | 123 | |||
XM_023653816.1 | XP_023509584.1 | 690 | 123 | |||
XM_023653817.1 | XP_023509585.1 | 690 | 123 | |||
XM_023653818.1 | XP_023509586.1 | 690 | 123 | |||
XM_023653819.1 | XP_023509587.1 | 690 | 123 | |||
XM_023653821.1 | XP_023509589.1 | 690 | 123 | |||
XM_023653824.1 | XP_023509592.1 | 690 | 123 | |||
GenBank mRNA | AB371895.1 | - | 4 - 5 | 658 | 123 | |
AB371896.1 | - | 4 - 5 | 658 | 123 | ||
AB371897.1 | - | 4 - 5 | 658 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |