Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101651.1 | NP_001095121.1 | 13 | 1646 | 125 | |
GenBank mRNA | EU014892.1 | - | 13 | 1646 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |