Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163879.1 | NP_001157351.1 | 2 - 3 | 249 | 102 | |
GenBank mRNA | AB035518.1 | - | 2 - 3 | 166 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |