Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163889.1 | NP_001157361.1 | 8 - 9 | 1561 | 62 | |
XM_005611061.3 | XP_005611118.1 | 1788 | 62 | |||
XM_023642149.1 | XP_023497917.1 | 2021 | 62 | |||
XM_023642150.1 | XP_023497918.1 | 1868 | 62 | |||
GenBank mRNA | AF076780.1 | - | 4 - 5 | 981 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Equus caballus Probe spans exons |