Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732990.2 | XP_014588476.2 | 604 | 76 | ||
XM_023638775.1 | XP_023494543.1 | 546 | 76 | |||
GenBank mRNA | AF359386.1 | - | 3 - 4 | 417 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |