Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163858.2 | NP_001157330.1 | 58 - 59 | 7957 | 71 | |
XM_023648324.1 | XP_023504092.1 | 8025 | 71 | |||
XM_023648325.1 | XP_023504093.1 | 8135 | 71 | |||
XM_023648326.1 | XP_023504094.1 | 7872 | 71 | |||
XM_023648328.1 | XP_023504096.1 | 8025 | 71 | |||
GenBank mRNA | AF448228.1 | - | 58 - 59 | 7874 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |