Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497655.6 | XP_001497705.5 | 1457 | 79 | ||
XM_023648954.1 | XP_023504722.1 | 924 | 79 | |||
XM_023648955.1 | XP_023504723.1 | 760 | 79 | |||
GenBank mRNA | AF506971.1 | - | 1 | 52 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |