Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655378.1 | XP_023511146.1 | 1138 | 71 | ||
GenBank mRNA | AF508034.1 | - | 4 - 5 | 677 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |