Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492151.6 | XP_001492201.6 | 783 | 81 | ||
XM_023643942.1 | XP_023499710.1 | 783 | 81 | |||
GenBank mRNA | AF510336.1 | - | 5 - 6 | 671 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |