Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145111.1 | NP_001138583.1 | 3 - 4 | 340 | 71 | |
XM_014740462.2 | XP_014595948.2 | 269 | 71 | |||
GenBank mRNA | AB472336.1 | - | 3 - 4 | 333 | 71 | |
AB477534.1 | - | 3 - 4 | 340 | 71 | ||
AY246702.1 | - | 3 - 4 | 292 | 71 | ||
HQ889955.1 | - | 3 - 4 | 301 | 71 | ||
JL616405.2 | - | 3 - 4 | 333 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Equus caballus Probe spans exons |