Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242479.1 | NP_001229408.1 | 4 - 5 | 2090 | 51 | |
GenBank mRNA | AY343542.1 | - | 2 - 3 | 361 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |