Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163953.1 | NP_001157425.1 | 6 - 7 | 1075 | 64 | |
XM_014728733.2 | XP_014584219.1 | 2050 | 64 | |||
XM_023633178.1 | XP_023488946.1 | 1409 | 64 | |||
XM_023633179.1 | XP_023488947.1 | 3485 | 64 | |||
GenBank mRNA | DQ367333.1 | - | 3 - 4 | 321 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |