Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163969.1 | NP_001157441.1 | 2 - 3 | 349 | 149 | |
GenBank mRNA | DQ681354.1 | - | 2 - 3 | 187 | 149 | |
HM015200.1 | - | 2 - 3 | 349 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Equus caballus Probe spans exons |