Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497744.2 | XP_001497794.1 | 1354 | 71 | ||
GenBank mRNA | EU029663.1 | - | 4 - 5 | 598 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Equus caballus Probe spans exons |