Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164013.2 | NP_001157485.2 | 7 - 8 | 662 | 107 | |
XM_023618413.1 | XP_023474181.1 | 948 | 107 | |||
XM_023618414.1 | XP_023474182.1 | 872 | 107 | |||
XM_023618416.1 | XP_023474184.1 | 877 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Equus caballus Probe spans exons |