Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164017.1 | NP_001157489.1 | 2 - 3 | 220 | 67 | |
XM_023625920.1 | XP_023481688.1 | 729 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |