Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAO2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242528.1 | NP_001229457.1 | 1 - 2 | 262 | 54 | |
XM_023636865.1 | XP_023492633.1 | 197 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |