Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245905.1 | NP_001232834.1 | 11 - 12 | 1475 | 61 | |
XM_023618423.1 | XP_023474191.1 | 1569 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |