Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245905.1 | NP_001232834.1 | 17 - 18 | 2120 | 112 | |
XM_023618423.1 | XP_023474191.1 | 2214 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |