Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170767.2 | NP_001164238.1 | 13 - 14 | 521 | 125 | |
XM_014738515.2 | XP_014594001.1 | 532 | 125 | |||
GenBank mRNA | GU196267.1 | - | 13 - 14 | 521 | 125 | |
KP658381.1 | - | 14 - 15 | 528 | 125 | ||
KP658382.1 | - | 12 - 13 | 477 | 125 | ||
KT368778.1 | - | 14 - 15 | 522 | 125 | ||
KT368779.1 | - | 12 - 13 | 471 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |