Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170767.2 | NP_001164238.1 | 4 - 5 | 151 | 102 | |
XM_014738515.2 | XP_014594001.1 | 111 | 102 | |||
GenBank mRNA | GU196267.1 | - | 4 - 5 | 151 | 102 | |
KP658381.1 | - | 3 - 4 | 107 | 102 | ||
KP658382.1 | - | 3 - 4 | 107 | 102 | ||
KT368778.1 | - | 3 - 4 | 101 | 102 | ||
KT368779.1 | - | 3 - 4 | 101 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |