Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185087.3 | NP_001172016.3 | 1 - 2 | 541 | 92 | |
NM_001185088.2 | NP_001172017.2 | 1 - 2 | 541 | 92 | ||
XM_005607780.3 | XP_005607837.1 | 630 | 92 | |||
XM_005607783.3 | XP_005607840.2 | 626 | 92 | |||
XM_023626768.1 | XP_023482536.1 | 628 | 92 | |||
XM_023626774.1 | XP_023482542.1 | 628 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Equus caballus Probe spans exons |