Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195635.1 | NP_001182564.1 | 7 - 8 | 1562 | 76 | |
XM_023651620.1 | XP_023507388.1 | 2684 | 76 | |||
GenBank mRNA | HM467836.1 | - | 7 - 8 | 1562 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |