Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195636.1 | NP_001182565.1 | 1 - 2 | 938 | 69 | |
XM_023638767.1 | XP_023494535.1 | 945 | 69 | |||
XM_023638768.1 | XP_023494536.1 | 945 | 69 | |||
XM_023638769.1 | XP_023494537.1 | 945 | 69 | |||
XM_023638770.1 | XP_023494538.1 | 945 | 69 | |||
GenBank mRNA | HM467835.1 | - | 1 - 2 | 938 | 69 | |
HM467837.1 | - | 1 - 2 | 938 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |