Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197094.1 | NP_001184023.1 | 1 - 2 | 197 | 68 | |
XM_005602134.3 | XP_005602191.1 | 513 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |