Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204845.1 | NP_001191774.1 | 7 - 8 | 1274 | 79 | |
XM_005599777.3 | XP_005599834.1 | 1239 | 79 | |||
XM_023618425.1 | XP_023474193.1 | 1192 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |