Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2D50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111306.1 | NP_001104776.1 | 6 | 921 | 63 | |
XM_023631011.1 | XP_023486779.1 | 1033 | 63 | |||
XM_023631012.1 | XP_023486780.1 | 1055 | 63 | |||
GenBank mRNA | EU190996.1 | - | 6 | 921 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |