Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204310.2 | NP_989641.2 | 7 - 8 | 1050 | 82 | |
XM_015283998.2 | XP_015139484.1 | 8 - 9 | 1601 | 82 | ||
XM_015283999.2 | XP_015139485.1 | 8 - 9 | 1600 | 82 | ||
XM_015284000.2 | XP_015139486.1 | 8 - 9 | 1595 | 82 | ||
XM_015284001.2 | XP_015139487.1 | 8 - 9 | 1578 | 82 | ||
XM_015284003.2 | XP_015139489.1 | 7 - 8 | 1521 | 82 | ||
GenBank mRNA | U86764.1 | - | 7 - 8 | 1050 | 82 | |
U86766.1 | - | 8 - 9 | 1077 | 82 | ||
U86767.1 | - | 8 - 9 | 1101 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |