Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277661.1 | NP_001264590.1 | 7 - 8 | 861 | 129 | |
NR_102417.2 | 6 - 7 | 667 | 129 | |||
XM_025148695.1 | XP_025004463.1 | 7 - 8 | 864 | 129 | ||
GenBank mRNA | BU111783.1 | - | 6 - 7 | 668 | 129 | |
BU371798.1 | - | 6 - 7 | 455 | 129 | ||
BU421743.1 | - | 4 - 5 | 329 | 129 | ||
BU441924.1 | - | 5 - 6 | 429 | 129 | ||
BX929417.1 | - | 7 - 8 | 862 | 129 | ||
CN383793.1 | - | 3 - 4 | 211 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |