Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424710.5 | XP_424710.2 | 2 - 3 | 636 | 88 | |
GenBank mRNA | BX929746.1 | - | 1 - 2 | 144 | 88 | |
BX933208.1 | - | 2 - 3 | 352 | 88 | ||
BX935761.1 | - | 2 - 3 | 273 | 88 | ||
CR386784.1 | - | 1 - 2 | 262 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |