Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285926.1 | NP_001272855.1 | 4 - 5 | 389 | 123 | |
XM_004938247.3 | XP_004938304.1 | 4 - 5 | 473 | 123 | ||
XM_004938248.3 | XP_004938305.1 | 4 - 5 | 389 | 123 | ||
GenBank mRNA | BI065382.1 | - | 3 - 4 | 232 | 123 | |
BX930121.1 | - | 4 - 5 | 386 | 123 | ||
BX930142.2 | - | 5 - 6 | 536 | 123 | ||
BX930156.1 | - | 5 - 6 | 536 | 123 | ||
BX930576.2 | - | 4 - 5 | 386 | 123 | ||
BX930906.1 | - | 4 - 5 | 399 | 123 | ||
BX931243.2 | - | 4 - 5 | 386 | 123 | ||
BX933264.1 | - | 4 - 5 | 369 | 123 | ||
BX935375.1 | - | 4 - 5 | 389 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |