Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199578.1 | NP_001186507.1 | 6 - 7 | 463 | 100 | |
XM_015281611.2 | XP_015137097.1 | 7 - 8 | 633 | 100 | ||
XM_015281618.2 | XP_015137104.1 | 6 - 7 | 533 | 100 | ||
XM_015281636.2 | XP_015137122.1 | 7 - 8 | 771 | 100 | ||
XM_015281639.2 | XP_015137125.1 | 7 - 8 | 596 | 100 | ||
XM_015281645.2 | XP_015137131.1 | 5 - 6 | 436 | 100 | ||
GenBank mRNA | BX930748.1 | - | 4 - 5 | 424 | 100 | |
BX933182.1 | - | 6 - 7 | 463 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |