Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277844.1 | NP_001264773.1 | 1 - 2 | 265 | 124 | |
NM_001277849.1 | NP_001264778.1 | 1 - 2 | 265 | 124 | ||
GenBank mRNA | BU317241.1 | - | 1 - 2 | 270 | 124 | |
BU405682.1 | - | 1 - 2 | 153 | 124 | ||
BX929747.1 | - | 1 - 2 | 144 | 124 | ||
BX931194.1 | - | 1 - 2 | 258 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |