Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942659.3 | XP_004942716.1 | 3 - 4 | 185 | 88 | |
GenBank mRNA | BX930573.1 | - | 2 - 3 | 172 | 88 | |
BX931475.1 | - | 2 - 3 | 165 | 88 | ||
BX932964.1 | - | 165 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |