Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171644.1 | NP_001165115.1 | 2 - 3 | 331 | 68 | |
NM_001171645.1 | NP_001165116.1 | 2 - 3 | 331 | 68 | ||
GenBank mRNA | BX929433.1 | - | 2 - 3 | 279 | 68 | |
BX929751.2 | - | 2 - 3 | 133 | 68 | ||
BX929814.1 | - | 2 - 3 | 290 | 68 | ||
BX931945.1 | - | 2 - 3 | 133 | 68 | ||
BX932880.1 | - | 2 - 3 | 271 | 68 | ||
BX933705.1 | - | 2 - 3 | 279 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |