Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271940.3 | NP_001258869.1 | 2 - 3 | 559 | 98 | |
NM_001271941.3 | NP_001258870.1 | 2 - 3 | 399 | 98 | ||
GenBank mRNA | BX932094.1 | - | 2 - 3 | 527 | 98 | |
BX932219.1 | - | 2 - 3 | 399 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Gallus gallus Probe spans exons |