Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197134.1 | NP_001184063.1 | 2 - 3 | 542 | 99 | |
NM_001197135.1 | NP_001184064.1 | 2 - 3 | 312 | 99 | ||
GenBank mRNA | BI390122.1 | - | 2 - 3 | 310 | 99 | |
BU300551.1 | - | 2 - 3 | 225 | 99 | ||
BX930146.2 | - | 2 - 3 | 266 | 99 | ||
BX930508.1 | - | 2 - 3 | 226 | 99 | ||
BX932215.1 | - | 2 - 3 | 257 | 99 | ||
BX932438.1 | - | 2 - 3 | 176 | 99 | ||
BX933260.2 | - | 2 - 3 | 547 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |