Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949575.3 | XP_004949632.1 | 4 - 5 | 540 | 99 | |
XM_015280691.2 | XP_015136177.1 | 4 - 5 | 452 | 99 | ||
XM_424940.6 | XP_424940.5 | 4 - 5 | 527 | 99 | ||
GenBank mRNA | BX932541.1 | - | 4 - 5 | 508 | 99 | |
BX934080.1 | - | 4 - 5 | 527 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |