Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282541.2 | XP_015138027.2 | 14 - 15 | 2105 | 101 | |
XM_015282543.2 | XP_015138029.2 | 14 - 15 | 2105 | 101 | ||
XM_025147818.1 | XP_025003586.1 | 14 - 15 | 2105 | 101 | ||
XM_025147819.1 | XP_025003587.1 | 13 - 14 | 1993 | 101 | ||
XM_025147820.1 | XP_025003588.1 | 14 - 15 | 2102 | 101 | ||
XM_025147821.1 | XP_025003589.1 | 10 - 11 | 1468 | 101 | ||
XM_025147824.1 | XP_025003592.1 | 14 - 15 | 2105 | 101 | ||
GenBank mRNA | BX933406.1 | - | 7 - 8 | 876 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |