Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277164.1 | NP_001264093.1 | 7 - 8 | 801 | 106 | |
GenBank mRNA | BX933609.1 | - | 6 - 7 | 787 | 106 | |
BX934117.1 | - | 5 - 6 | 538 | 106 | ||
BX934597.2 | - | 6 - 7 | 787 | 106 | ||
CR386179.1 | - | 7 - 8 | 607 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |