Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282335.2 | XP_015137821.1 | 1 - 2 | 149 | 110 | |
XM_419061.5 | XP_419061.1 | 1 - 2 | 159 | 110 | ||
GenBank mRNA | BX929367.1 | - | 1 - 2 | 127 | 110 | |
BX933021.1 | - | 1 - 2 | 149 | 110 | ||
BX933813.1 | - | 1 - 2 | 149 | 110 | ||
BX934156.1 | - | 1 - 2 | 127 | 110 | ||
BX934371.1 | - | 1 - 2 | 138 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |