Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM189A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025144867.1 | XP_025000635.1 | 4 - 5 | 431 | 64 | |
XM_025144868.1 | XP_025000636.1 | 4 - 5 | 383 | 64 | ||
XM_424828.6 | XP_424828.4 | 4 - 5 | 699 | 64 | ||
GenBank mRNA | BX933855.1 | - | 2 - 3 | 172 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |