Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199430.1 | NP_001186359.1 | 7 - 8 | 622 | 69 | |
GenBank mRNA | BX933953.1 | - | 6 - 7 | 445 | 69 | |
CR386813.1 | - | 1 - 2 | 101 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |