Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277999.1 | NP_001264928.1 | 3 - 4 | 314 | 83 | |
XM_015285101.2 | XP_015140587.1 | 4 - 5 | 344 | 83 | ||
XM_015285102.2 | XP_015140588.1 | 4 - 5 | 322 | 83 | ||
GenBank mRNA | BX930018.1 | - | 3 - 4 | 326 | 83 | |
BX934024.1 | - | 3 - 4 | 314 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |