Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX934167.1 | - | 2 - 3 | 138 | 85 | |
BX934174.2 | - | 2 - 3 | 138 | 85 | ||
BX934237.1 | - | 2 - 3 | 138 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |