Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277520.1 | NP_001264449.1 | 2 - 3 | 213 | 85 | |
NM_001277521.1 | NP_001264450.1 | 2 - 3 | 213 | 85 | ||
GenBank mRNA | BU375910.1 | - | 2 - 3 | 213 | 85 | |
BX929409.2 | - | 1 - 2 | 53 | 85 | ||
BX934691.1 | - | 2 - 3 | 209 | 85 | ||
CV854591.1 | - | 2 - 3 | 193 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |