Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414580.5 | XP_414580.1 | 12 - 13 | 1503 | 108 | |
GenBank mRNA | BX934896.1 | - | 9 - 10 | 921 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |