Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942397.3 | XP_004942454.1 | 5 - 6 | 1323 | 152 | |
XM_004942398.3 | XP_004942455.1 | 5 - 6 | 1323 | 152 | ||
XM_004942399.3 | XP_004942456.1 | 4 - 5 | 1077 | 152 | ||
XM_025152269.1 | XP_025008037.1 | 5 - 6 | 1323 | 152 | ||
GenBank mRNA | BX930694.2 | - | 3 - 4 | 530 | 152 | |
BX930963.2 | - | 3 - 4 | 1228 | 152 | ||
BX934991.1 | - | 5 - 6 | 1107 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |