Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277518.1 | NP_001264447.1 | 5 - 6 | 1314 | 73 | |
XM_015283466.2 | XP_015138952.1 | 5 - 6 | 1115 | 73 | ||
XM_015283467.2 | XP_015138953.1 | 5 - 6 | 1342 | 73 | ||
XM_015283468.2 | XP_015138954.1 | 5 - 6 | 1314 | 73 | ||
GenBank mRNA | BX929403.2 | - | 5 - 6 | 1299 | 73 | |
BX929927.2 | - | 5 - 6 | 1069 | 73 | ||
DR415418.1 | - | 801 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |